(7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methanol

Supplier: Matrix Scientific

070900-250MG
102519-088EA 709.23 USD
102519-088
(7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methanol
(7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridin-8-yl)methanol

Hazard: Irritant

Formula: C₈H₈ClNO₃
MDL Number: MFCD20487092
CAS Number: 1346447-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR