7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime
Supplier: Matrix Scientific
070894-250MG
102519-076EA
860
USD
102519-076
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde oxime
Hazard: Irritant
Formula:
C₈H₇ClN₂O₃ MW: 214.61 g/mol |
MDL Number:
MFCD20487087 CAS Number: 1346451-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...