7-Chloro-8-(trimethylsilyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

070892-250MG
102519-072EA 709.23 USD
102519-072
7-Chloro-8-(trimethylsilyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
7-Chloro-8-(trimethylsilyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂Si
MW: 243.77 g/mol
MDL Number: MFCD20487084
CAS Number: 1305324-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR