7,8-Dichloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

070888-250MG
102519-064EA 709.23 USD
102519-064
7,8-Dichloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
7,8-Dichloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
MDL Number: MFCD20487076
CAS Number: 1305324-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR