7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde

Supplier: Matrix Scientific

070887-250MG
102519-062EA 766.15 USD
102519-062
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde
7-Chloro-2,3-dihydro[1,4]dioxino[2,3-b]pyridine-8-carbaldehyde

Hazard: Irritant

Formula: C₈H₆ClNO₃
MW: 199.59 g/mol
MDL Number: MFCD20487075
CAS Number: 1305324-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR