3,4-Dihydro-2H-pyrano[3,2-b]pyridine-8-carbaldehyde

Supplier: Matrix Scientific

070881-250MG
102519-050EA 481.54 USD
102519-050
3,4-Dihydro-2H-pyrano[3,2-b]pyridine-8-carbaldehyde
3,4-Dihydro-2H-pyrano[3,2-b]pyridine-8-carbaldehyde

Hazard: Irritant

Formula: C₉H₉NO₂
MDL Number: MFCD20487143
CAS Number: 1346446-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR