6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridin-8-ol

Supplier: Matrix Scientific

070876-250MG
102519-040EA 726.15 USD
102519-040
6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridin-8-ol
6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridin-8-ol

Hazard: Irritant

Formula: C₈H₈INO₂
MDL Number: MFCD20487127
CAS Number: 1346447-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR