6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine-8-carbonitrile

Supplier: Matrix Scientific

070874-250MG
102519-036EA 860 USD
102519-036
6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine-8-carbonitrile
6-Iodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine-8-carbonitrile

Hazard: Irritant

Formula: C₉H₇IN₂O
MW: 286.07 g/mol
MDL Number: MFCD20487125
CAS Number: 1346447-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR