(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)methanol
Supplier: Matrix Scientific
070873-250MG
102519-034EA
709.23
USD
102519-034
(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)methanol
(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)methanol
Hazard: Irritant
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD20487110 CAS Number: 1346446-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...