N-(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)pivalamide
Supplier: Matrix Scientific
070870-250MG
102519-028EA
526.15
USD
102519-028
N-(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)pivalamide
N-(3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-yl)pivalamide
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₂ MW: 234.3 g/mol |
MDL Number:
MFCD20487107 CAS Number: 1346447-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...