3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine

Supplier: Matrix Scientific

070866-250MG
102519-020EA 526.15 USD
102519-020
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine

Hazard: Irritant

Formula: C₈H₁₀N₂O
MDL Number: MFCD20487103
CAS Number: 1346447-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR