2-Phenyl-4-(trifluoromethyl)quinoline ≥97%
Supplier: Matrix Scientific
070847-1G
102518-962EA
186.15
USD
102518-962
2-Phenyl-4-(trifluoromethyl)quinoline ≥97%
2-Phenyl-4-(trifluoromethyl)quinoline
Hazard: Irritant
Formula:
C₁₆H₁₀F₃N Melting Pt: 37…39 °C |
MDL Number:
MFCD11226599 CAS Number: 64196-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...