2-Bromo-5-(1,1-difluoroethyl)pyridine ≥95%

070846-500MG
102518-960EA 253.49 USD
102518-960
2-Bromo-5-(1,1-difluoroethyl)pyridine ≥95%
2-Bromo-5-(1,1-difluoroethyl)pyridine

Hazard: Irritant

Formula: C₇H₆BrF₂N
Boiling Pt: 80 °C
MDL Number: MFCD16140182
CAS Number: 1211521-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR