Rat Neural Stem Cells (RNSC) are isolated from cerebral striatum and mesencephalon of E18 fetal rat brains.
- Maintained in undifferentiated proliferative state
- Cryopreserved at second passage
- Multipotent and can be induced to differentiate into neurons
Rat Neural Stem Cells (RNSC) are isolated from cerebral striatum and mesencephalon of E18 fetal rat brains. These cells were maintained in undifferentiated proliferative state by culturing them as free floating neurospheres in serum-free medium optimized with growth factors. They are cryopreserved at second passage and can be cultured and propagated for one to two passages prior to induction of phenotypic differentiation. These stem cells are multipotent and can be induced to differentiate into neurons, astrocytes and oligodendrocytes that make up the central nervous system (CNS). Neurospheres transplanted into intact brain can survive, expand and differentiate into mature neurons, astrocytes and oligodendrocytes precursors in three weeks. The ability of neural stem cells to retain multi-lineage potential and proliferate extensively in vitro provides new avenues for the treatment of neural degenerative diseases and injuries.