BBL™ Transport Medium, BD Biosciences
Supplier: BD Biosciences
211743 211743
90000-316EA 461.06 USD
CA90000-316 90000-316
BBL™ Transport Medium, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
BBL™ Transport Medium is a semisolid medium recommended for use in the transport and preservation of microbiological specimens.
  • Non-nutritional medium
  • Maintains the viability of organisms without significant multiplication
  • Small agar content provides a semisolid consistency
  • Prevents oxidation and drying during transport
Transport media are designed to preserve the viability of microorganisms when immediate inoculation of growth media is not possible.