Tegaderm™ Transparent Film Dressing with Border, 3M™
Supplier: 3M
Tegaderm™
MMM1655 MMM1650 MMM1610 MMM1633 MMM1635
75787-456CS 1035.73 USD
75787-456 75787-458 75787-810 75788-420 75787-808
Tegaderm™ Transparent Film Dressing with Border, 3M™
First Aid Supplies Wound Dressings
Transparent I.V dressing reinforced with a soft cloth fabric border, designed to seal around the insertion site and minimize catheter movement.
  • Transparency allows monitoring of I.V. sites without the need to remove the dressing
  • Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria, and viruses
  • Seven days wear time