Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Wall-Mounted Hose Racks, Integra Companies

Supplier: INTEGRA COMPANIES
HSU-24 HSU-11 HSU-32 HSU-MINI HSU-100 HSU-16
The arch and slot style hose racks accommodate a variety of hoses, and are available in standard or mini sizes. These wall-mounted racks keep various size hoses organized and off the floor.

  • Extends hose working life and ensures 100% drainage
  • Compact designs ideal for use in any cleanroom enviornment
  • Eliminates clutter and enhances safety
  • Convieniently stores sanitary gaskets, clamps, and adaptors in one location (not included on Arch Rack Mini)
  • Constructed from 304 electropolished stainless steel
  • Ideal for biopharmaceutical cleanroom applications

Three unique wall-mounted hose rack styles are available to choose from, including slot, arch, and mini-arch shaped designs.

Certifications: Meets FDA and 3A regulations for hose storage, arch and slot style racks suits cGMP.

Delivery information: Includes mounting hardware.



 

ORDER

Expand All / Collapse All
Description Dimensions Details Supplier No. VWR Catalog Number Unit Price Quantity
Arch Style Hose Rack 15.2W×40.6H×45.7L cm (6×16×18") Holds a Variety of Hoses HSU-100 10014-873 Each Retrieving
Mini Arch Style Hose Rack 10.2W×28H×30.5L cm (4×11×12") Holds a Variety of Small Length Hoses HSU-MINI 10014-875 Each Retrieving
Slot Style Hose Rack 13W×15.2H×15.2L cm (5.13×6× 6") Holds 12 Hoses of 1.9 cm (0.75") each HSU-11 10014-877 Each Retrieving
Slot Style Hose Rack 13W×15.2H×15.2L cm (5.13×6×6") Holds 4 Hoses of 2.5 cm (1") each HSU-16 10014-879 Each Retrieving
Slot Style Hose Rack 13W×15.2H×15.2L cm (5.13×6×6") Holds 4 Hoses of 3.8 cm (1.5") each HSU-24 10014-881 Each Retrieving
Slot Style Hose Rack 14.3W×15.2H×15.2L cm (5.63×6×6") Holds 5 Hoses of 5 cm (2") each HSU-32 10014-883 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty