Phytone™ Peptone, Life Technologies
Supplier: Life Technologies
215263
71002-198EA 876.19 USD
71002-198
Phytone™ Peptone, Life Technologies
Phytone peptone is an enzymatic digest of soybean meal, designed specifically for cell culture applications.
  • Non-animal origin
  • Cultivates a wide variety of organisms
  • Contains high concentrations of vitamins and carbohydrates
Recommended for use in media for the cultivation of a wide variety of organisms, including fungi. Phytone™ peptone was developed specifically for the tissue culture market, with its nitrogen content combined with naturally occurring vitamins demonstrating remarkable growth support.