PrimaPure™, Rat Aortic Smooth Muscle Cell Culture Kit (RAOSMC), Genlantis
Supplier: Genlantis
SR35405K SR35405
10497-912EA 1765.84 USD
10497-912 10497-910
PrimaPure™, Rat Aortic Smooth Muscle Cell Culture Kit (RAOSMC), Genlantis
Cells
PrimaPure™ Rat Aortic Smooth Muscle Cells (RAOSMC) are derived from the tunica intima and tunica media of healthy, fibrous plaque-free rat aorta. Increased arterial smooth muscle cell mass are found in the intimal lesion of atherosclerosis.
  • Each lot functionally characterized
  • Complete kits or cells only
  • Cryopreserved at second passage
  • Tested negative for mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated at least 8 population doublings
Rat aortic smooth muscle cells respond to various factors by cell proliferation and hypertrophy, which are prominent indicators of atherosclerosis in vascular diseases
Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.
RAOSMC are well suited for the study of large vessel smooth muscle cell growth and differentiation4,5 and serve as in vitro model in correlation with live rat models.
Delivery information: Kit contains an ampoule of cryopreserved RAOSMC, 500 mL of Rat Smooth Muscle Cell Growth Medium, and a Subculture Reagent Kit.