Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Rat Cardiac Fibroblasts Cell Culture Kit (RCF), Genlantis

Supplier: Genlantis
SR30605A SR30605AK
PrimaPure™ Rat Cardiac Fibroblasts (RCF) are derived from the heart tissue of normal Sprague Dawley rats. Rat Cardiac Fibroblasts are known to play key roles in the development of cardiac disease and hypertrophy.

  • Each lot functionally characterized
  • Complete kits or cells only
  • Cryopreserved at second passage
  • Tested negative for mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated at least 8 population doublings

The regulation of fibroblast growth after acute myocardial infarction was determined in RCF

RCF have also been used to study the autocrine/paracrine capability mediated by vasodilator such as adernomedullin. RCF are used to investigate their role in local hormone metabolism. Nitric oxide (NO) production in RCF decreases cAMP accumulation largely by the cGMP-mediated activation of PDE2. IL-1 beta-induced inducible NO synthase expression can trigger NO-dependent apoptosis in adult RCF.

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Cardiac fibroblasts were examined as a potential source of pro-inflammatory cytokine IL-6 synthesis which had been implicated in the pathophysiology of heart disease.

Delivery information: Each kit contains an ampoule of cryopreserved RCF, 500 mL of Rat Fibroblast Growth Medium, and a Subculture Reagent Kit.



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Rat Cardiac PrimaPure™: Normal Cardiac Fibroblasts Cells >500,000 Cryopreserved SR30605A 10497-906 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Cardiac PrimaPure™: Normal Cardiac Fibroblasts Cell, Complete System Kit - SR30605AK 10497-908 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Rat PrimaPure™ Fibroblast Basal Medium 1 x 500 mL PMR115500 10497-428 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat PrimaPure™ Fibroblast Growth Medium 1 x 500 mL PMR116500 10497-430 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat PrimaPure™ Fibroblast Growth Supplement 1 x 50 mL PMR116GS 10497-432 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty