Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Rat Aortic Endothelial Cell Culture Kit (RAOEC), Genlantis

Supplier: Genlantis
SR30405 SR30405K
Rat Aortic Endothelial Cells (RAOEC) are isolated from aorta of healthy adult rats.

  • Each lot functionally characterized
  • Complete kits or cells only
  • Cryopreserved at third passage
  • Tested negative for mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated to sixth passage and beyond without losing their morphologic and phenotypic characteristics

RAOEC is a relevant in vitro model for correlating with in vivo studies in rat as an animal model

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

RAOEC have been used as a model for the study of atherosclerosis and products synthesized by the endothelium. The interaction of aortic endothelium and smooth muscle cells regulate the level and distribution of glycosaminoglycans in the arterial wall, especially in the intima. Co-culture of aortic endothelial cells and smooth muscle cells provide an ideal in vitro model for studying the interaction between these two cells types.

Delivery information: Kit contains an ampoule of cryopreserved RAOEC, 500 mL of Rat Endothelial Cell Growth Medium, 100 mL of Attachment Factor Solution and a Subculture Reagent Kit.



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Rat Aorta PrimaPure™: Normal Aortic Endothelial Cells >500,000 Cryopreserved SR30405 10497-902 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Aorta PrimaPure™: Normal Aortic Endothelial Cells, Complete System Kit - SR30405K 10497-904 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Rat PrimaPure™ Endothelial Cell Basal Medium 1 x 500 mL PMR210500 10497-434 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat PrimaPure™ Endothelial Cell Growth Medium 1 x 500 mL PMR211500 10497-436 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat PrimaPure™ Endothelial Cell Growth Supplement 1 x 15 mL PMR211GS 10497-438 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty