Carbenicillin disodium salt, powder, Ultrapure

Supplier: Genlantis

Synonyms: 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Naα-Carboxybenzylpenicillin disodium salt

M110250 M111000
10497-864EA 228.7 USD
10497-864 10497-862
Carbenicillin disodium salt, powder, Ultrapure
Carbenicillin disodium salt

A penicillin derivative that inhibits cell wall synthesis in Gram-negative and Gram-positive bacteria


Working concentration: 0.1-30 µg/mL.

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
CAS Number: 4800-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR