Hygromycin B, ultrapure
M140100
10497-872EA 131.24 USD
10497-872
Hygromycin B, ultrapure
Hygromycin B
An aminoglycoside antibiotic that inhibits protein synthesis in bacteria, fungi and higher eucaryotic cells.
  • Widely used as a selection agent
  • An aminoglycoside antibiotic
  • Inhibits protein synthesis
An aminoglycoside antibiotic that inhibits protein synthesis in bacteria, fungi and higher eucaryotic cells. Widely used as a selection agent for procaryotic and eucaryotic cells carrying the hygromycin resistance gene. Working concentration: 100 µg/mL.