PerFectin™ Transfection Reagent, Genlantis
Supplier: Genlantis
PerFectin™
T303007 T303015
10497-818EA 336.93 USD
10497-818 10497-820
PerFectin™ Transfection Reagent, Genlantis
Transfection Reagents
PerFectin™ Transfection Reagent is a unique formulation of the neutral lipid, dioleoyl phosphatidylethanolamine (DOPE) and a proprietary cationic lipid derived from Direct Hydrophilic Conjugation (DHC) technology
  • High efficiency with CHO, COS, 293, HeLa, NIH-3T3, and Jurkat cells
  • Extended shelf life with superior transfection efficiency
  • Easy and optimized protocols for individual cell lines
  • Compatible with serum containing media
This reagent is suitable for both in vitro and in vivo transfection studies
Delivery information: The kit is supplied with a special DNA Diluent that increases transfection efficiency in the presence of serum.