Would you like to log in again?
PrimaPure™ Rat Marrow Stromal Cells (RMSC), Genlantis
Supplier: Genlantis
SR49205 SR49205K
Rat Marrow Stromal Cells (RMSC)/Rat Mesenchymal Stem Cells (MSC) are isolated from rat bone marrow and are ideal for investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells.
  • Cryopreserved at second passage
  • Tested negative for mycoplasma, bacteria, yeast, and fungi
  • Each lot is functionally characterized
  • can be cultured and propagated 10 population doublings
RMSC have the ability to self-renew and differentiate into cells of various connective tissue lineages, such as bone, cartilage, fat, and neurons
Each lot of PrimaPure MSC's is tested for its ability to attach and spread on standard tissue culture grade plastic surfaces and proliferate in Rat Marrow Stromal Cell Growth Medium. Additionally, each lot is characterized for bone mineralization and lipid accumulation following differentiation using osteoblast and adipocyte differentiation media, repectively.
Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.
The potential of MSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.
Delivery information: Kit contains an ampoule of cryopreserved RMSC, 500 mL of Rat Marrow Stromal Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, Trypsin/EDTA, and Trypsin Neutralizing Solution).
ORDER
Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
PrimaPure™ Normal Rat Marrow Stromal Cells >500,000 Cryopreserved SR49205 10497-734 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PrimaPure™ Normal Rat Marrow Stromal Cells, Complete System Kit Cryopreserved SR49205K 10497-736 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty