PrimaPure™, Mouse Neural Stem Cells (MNSC), Genlantis
Supplier: Genlantis
SM82020F SM82020FK
10497-726EA 2551.88 USD
10497-726 10497-724
PrimaPure™, Mouse Neural Stem Cells (MNSC), Genlantis
Cells
Mouse Neural Stem Cells (MNSC) are isolated from cerebral striatum and mesencephalon of E14 fetal mouse brains.
  • Each lot functionally characterized
  • Cryopreserved at second passage
  • Negative for mycoplasma, bacteria, yeast, and fungi
  • Can be can be cultured and propagated for 1- 2 passages prior to induction of phenotypic differentiation
These cells are maintained in undifferentiated proliferative state by culturing them as free floating neurospheres in serum-free medium optimized with growth factors
Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.
Stem cells are multipotent and can be induced to differentiate into neurons, astrocytes and oligodendrocytes. Neural stem cells are a source of donor tissue for prospective neural cell therapy research given the ability of neurosphere to retain multi-lineage potential and proliferative ability in vitro.
Ordering information: Mouse Neural Stem Cell Differentiation Medium and the Poly-D-Lysine with Laminin Coating Solution are available separately on request.
Delivery information: Each kit contains an ampoule of cryopreserved MNSC, 250 mL of Mouse Neural Stem Cell Growth Medium and two non-tissue culture treated 10 cm plates.