Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Porcine Aortic Smooth Muscle Cell Culture Kit (PAOSMC), Genlantis

Supplier: Genlantis
PP35405 PP35405K
Porcine large artery smooth muscle cells are derived from tunica intima and tunica media of healthy, fibrous plaque-free porcine arteries.

  • Positive for expression of smooth muscle cell specific α-actin
  • Negative for mycoplasma, bacteria, yeast, and fungi
  • Cryopreserved at second passage
  • Can be cultured and propagated at least 14 population doublings

Porcine Aortic Smooth Muscle Cells (PAOSMC) serve as an alternative for the investigations of cardiac disease and arteriosclerosis

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

It can be used in co-culture investigation with species-matched porcine artery endothelial cells.

Delivery information: Kit contains an ampoule of cryopreserved PAOSMC, 500 mL of Porcine Smooth Muscle Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Porcine Artery PrimaPure™: Normal Aortic Smooth Muscle Cells >500,000 Cryopreserved PP35405 10497-722 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Porcine Artery PrimaPure™: Normal Aortic Smooth Muscle Cells, Complete System Kit - PP35405K 10497-444 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Porcine PrimaPure™ Smooth Muscle Cell Growth Medium 1 x 500 mL PMP311500 10497-426 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty