Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DRY-KEEPER™ Dessicator Cabinet, Chemglass

Supplier: Chemglass
CG-1225-11
These desiccator cabinets molded of methyl-methacrylate offer a clear view of the cabinet contents

  • Cabinet includes 3 sets of shelf rails and 3 clear shelves with small vent holes
  • The gasketed door has two metal hinges, two latches, and a handle
  • Door opening is 10” wide x 17 7/8” high
  • Inside cabinet depth is 13 1/4
  • The overall dimensions are 12 1/2” wide x 20” high x 15 1/4” deep

Cabinet comes complete with a dial hygrometer and desiccant tray.This is equipped with a 110V, 6 watt regeneration unit for the silica gel. The gel is contained in the control section, where it absorbs moisture to maintain a low (30-40%) relative humidity. The absorption period lasts 4 1/2 hours, indicated by a green pilot light. After this period, the silica gel is regenerated for a 1/2 hour period, indicated by a red pilot light. After regeneration of the silica gel, this cycle is repeated. The electric regeneration unit adds 6 inches to the width of the cabinet.

Cleats allow for 15 levels of shelf placement.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Dry keeper, Desiccator Cabinets CG-1225-11 80060-656 Each Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Overall Dimensions 50.8H x 31.75W x 39.37D cm (20 x 12 1/2 x 15 1/4")

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty