Fibroblast Growth Medium, Genlantis
Supplier: Genlantis
PM116500 PM115500
10497-396EA 319.56 USD
10497-396 10497-394
Fibroblast Growth Medium, Genlantis
Cell Culture Media
Fibroblasts are cells that make the structural fibers and ground substance of connective tissue.
  • Ground substance of connected tissue
  • Branched cytoplasm
  • Make collagens, glycosaminoglycans, reticular and elastic fibers
Fibroblasts have a branched cytoplasm surrounding an elliptical, speckled nucleus having 1 or 2 nucleoli. Fibroblasts make collagens, glycosaminoglycans, reticular and elastic fibers, and glycoproteins found in the extracellular matrix. In growing individuals fibroblasts are dividing and synthesizing ground substance. Tissue damage stimulates fibrocytes and induces the mitosis of fibroblasts.