PrimaPure™, Bovine Aortic Smooth Muscle Cell Culture Kit (BAOSMC), Genlantis
Supplier: Genlantis
PB35405K PB35405
10497-352EA 1431.18 USD
10497-352 10497-350
PrimaPure™, Bovine Aortic Smooth Muscle Cell Culture Kit (BAOSMC), Genlantis
Cells
Bovine aortic smooth muscle cells (BAOSMC) are derived from tunica intima and tunica media of healthy, fibrous, plaque-free bovine large aorta.
  • Each lot functionally characterized
  • Same quality standard as the PrimaPure human cell products
  • Tested negative for mycoplasma, bacteria, yeast and fungi
  • Cryopreserved at the second passage
  • Can be cultured and propagated at least 16 population doublings
Bovine smooth muscle cells are an economical alternative suitable for investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.
Proliferation of the smooth muscle cells is considered a key event in the development of advanced lesions.
Delivery information: Each complete system contains an ampoule of cryopreserved BAOSMC (> 500,000 cells), 500 mL Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution).