Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Bovine Coronary Artery Smooth Muscle Cell Culture Kit (BCASMC), Genlantis

Supplier: Genlantis
PB35005 PB35005K
Bovine coronary artery smooth muscle cells (BCASMC) are derived from tunica intima and tunica media of healthy, fibrous, plaque-free bovine large arteries.

  • Each lot functionally characterized
  • Same quality standard as the PrimaPure human cell products
  • Tested negative for mycoplasma, bacteria, yeast and fungi
  • Can be cultured and propagated at least 16 population doublings

Bovine smooth muscle cells are an economical alternative suitable for investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Proliferation of the smooth muscle cells is considered a key event in the development of advanced lesions.

Delivery information: Each kit contains an ampoule of cryopreserved BCASMC, 500 mL of Bovine Smooth Muscle Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Bovine Artery PrimaPure™: Normal Coronary Artery Smooth Muscle Cells >500,000 cells Cryopreserved PB35005 10497-342 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Bovine Artery PrimaPure™: Normal Coronary Artery Smooth Muscle Cells, Complete System Kit - PB35005K 10497-344 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Bovine PrimaPure™ Smooth Muscle Cell Growth Medium 1 x 500 mL PMB311500 10497-422 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty