Kanamycin sulfate, ultrapure

Synonyms: Kanamycin MonosulphateKanamycin AKanamycin Monosulfate

M150025 M150010
10497-318EA 355.95 USD
10497-318 10497-316
Kanamycin sulfate, ultrapure
Kanamycin sulphate

Kanamycin is an aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma.


  • Aminoglycoside antibiotic
  • Optimized for cell culture


Kanamycin is an aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma. Working concentration: 30 µg/mL. These antibiotics are optimized for cell culture.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR