Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Detachin™ Cell Detachment Solution, Genlantis

Supplier: Genlantis
T100110 T100100 T100106
Detachin™ Cell Detachment Solution is a superior alternative to Trypsin/EDTA for gently detaching adherent cells from in vitro growth vessels. Detachin™ provides quick, gentle, and effective detachment of a wide variety of adherent cells, including primary cells, from all known tissue culture plastic ware. It contains protease and collagenase activities in an isotonic, phosphate buffer solution with EDTA.

  • Gentle and rapid detachment with maximum cell viability
  • Effective on a wide range of cells
  • No mammalian or bacterial by-products
  • Stable at 4 °C for two months
  • No need to wash detached cells
  • Convenient and formats including 100 mL, 10 x 100 mL, and 6-PAC (6 x 50 mL) sizes

Detachin™ Cell Detachment Solution is a superior alternative to Trypsin/EDTA for gently detaching adherent cells from in vitro growth vessels

Available in different convenient formats including 100 mL, 10 x 100 mL, and 6-PAC (6 x 50 mL) sizes. The Detachin 6-PAC format allows users to preserve unused portions of Detachin for the long term, reduce the chance of contamination of Detachin stock, and reduce footprint of Detachin in tissue culture hoods.

Detachin™ helps to get a novel solution that provides consistent, safe, and efficient results. It has been tested successfully on a wide variety of different cells and cell types like bone marrow stem, fibroblasts, hepatocytes, mouse germ, keratinocytes, A-375, BHK, CHO, COS , D54, HEK 293, HeLa, and many others.

Detachin™ provides quick, gentle, and effective detachment of a wide variety of adherent cells, including primary cells, from all known tissue culture plastic ware. It contains protease and collagenase activities in an isotonic, phosphate buffer solution with EDTA.



 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Detachin™ Cell Detachment Solution 100 mL bottle T100100 10497-740 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Detachin™ Cell Detachment Solution, 6-PAC 6 x 50 mL tubes T100106 10497-742 Pack of 6 Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Detachin™ Cell Detachment Solution 10 x 100 mL bottles T100110 10497-744 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty