Swift Advanced Stereo Zoom Microscope
Swift®
M29TZ-SM99CL
470222-612EA 929.8 USD
470222-612
Swift Advanced Stereo Zoom Microscope
Microscopes
Ideal for educational, laboratory, research and industrial use.
  • Viewing head rotates 360 degrees
  • 80mm working distance
  • Top and bottom LED illumination
  • C-mount port for easy addition of optional video C-mount and SLR cameras
This stereo zoom microscope offers the convenience and versatility of viewing items at 10X magnification then zoom continuously to maximum detail at 40X magnification or at any point in between. The upright, unreversed image remains in focus throughout the zoom range. The trinocular head features a port to accept optional video C-mount and SLR camera adapters' included.
Note: the incidental and transmitted illuminators incorporate high intensity 3W LED lamps.