Low Range DNA Molecular Weight Marker, Genlantis
D402100
10497-304EA 143.13 USD
10497-304
Low Range DNA Molecular Weight Marker, Genlantis
High quality DNA molecular weight marker for a variety of electrophoretic applications.
  • Idela for use in electrophoresis of DNA in agarose and polyacrylamide gels.
  • Ideal for determining the size of a wide range of DNA fragments
  • Sensitive and quick staining of DNA
The low range DNA marker I contains 14 DNA bands ranging in size from 8.4 kb to 0.1 kb.