PrimaPure™, Human Aortic Smooth Muscle Cell Culture Kit (HAOSMC), Genlantis

Supplier: Genlantis
PH35405A PH35405AK
10497-672EA 1187.05 USD
10497-672 10497-674
PrimaPure™, Human Aortic Smooth Muscle Cell Culture Kit (HAOSMC), Genlantis
Cells
The PrimaPure™ system has been developed for the acquisition and growth of primary cells which are derived from the tunica intima and tunica media of normal human, fibrous plaquefree aorta.

  • Characterized positive for expression of smooth muscle cell specific α-actin
  • Tested negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated at least 16 population doublings
  • Lots from single donors to allow variability comparisons
  • Cryopreserved at second passage

HAOSMC respond to various factors by proliferating or differentiating

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

They are a well established cell system for the study of human vascular disorders such as atherosclerosis and stroke.

Delivery information: Kit contains an ampoule of cryopreserved HAOSMC, 500 mL of Smooth Muscle Cell Growth Medium and a Subculture Reagent Kit (including 100 ml each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR