Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Human Coronary Artery Smooth Muscle Cell Culture Kit (HCASMC), Genlantis

Supplier: Genlantis
PH35005A PH35005AK
The PrimaPure™ system has been developed for the acquisition and growth of primary cells which are derived from the tunica intima and tunica media of normal human, fibrous plaquefree coronary arteries.

  • Characterized positive for expression of smooth muscle cell specific α-actin
  • Tested negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated at least 16 population doublings
  • Lots from single donors to allow variability comparisons
  • Cryopreserved at second passage

HCASMC is an ideal cellular model for the investigations of coronary artery diseases, such as alteration of smooth muscle cell differentiation in atherosclerosis.

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Delivery information: Kit contains an ampoule of cryopreserved HCASMC, 500 mL of Smooth Muscle Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Human Artery PrimaPure™: Normal Adult Coronary Artery Smooth Muscle Cells >500,000 Cryopreserved PH35005A 10497-664 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Artery PrimaPure™: Normal Adult Coronary Artery Smooth Muscle Cell, Complete System Kit - PH35005AK 10497-666 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Human PrimaPure™ Smooth Muscle Cell Basal Medium 1 x 500 mL PM310500 10497-684 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Smooth Muscle Cell Growth Medium 1 x 500 mL PM311500 10497-686 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Smooth Muscle Cell Differentiation Medium 1 x 250 mL PM311D250 10497-688 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Smooth Muscle Cell Serum free Growth Medium 1 x 500 mL PM311F500 10497-690 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Smooth Muscle Cell Growth Supplements 1 x 30 mL PM311GS 10497-692 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty