Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Human Pulmonary Artery Endothelial Cell Culture Kit (HPAEC), Genlantis

Supplier: Genlantis
PH30205AK PH30205A
The PrimaPure™ system has been developed for the acquisition and growth of primary cells isolated from normal human pulmonary arteries. Human Pulmonary Artery Endothelial Cell (HPAEC) respond to a wide range of vasoactive substances, commensurate with their control of blood pressure and blood pH in vivo.

  • Von Willebrand factor/factor VIII (vWF) positive
  • Negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated at least 15 population doublings
  • Single donors to allow variability comparisons
  • Cryopreserved at second passage
  • Possess an array of enzymatic activities

The system is commonly used for the study of vascular permeability and inflammatory responses

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

HPAEC in co-culture with HPASMC have been used as a model for pulmonary angiopathy.

Delivery information: Kit contains an ampoule of cryopreserved HPAEC, 500 mL of Endothelial Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Human Lung PrimaPure™ : Normal Adult Pulmonary Artery Endothelial Cells >500,000 Cryopreserved PH30205A 10497-656 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Lung PrimaPure™ : Normal Adult Pulmonary Artery Endothelial Cell, Complete System Kit - PH30205AK 10497-658 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Human PrimaPure™ Endothelial Cell Serum-Free Defined Medium 1 x 500 mL PM113500 10497-390 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Endothelial Cell Serum-Free, Growth Factor-Free Medium 1 x 500 mL PM114500 10497-392 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Endothelial Cell Basal Medium 1 x 500 mL PM210500 10497-408 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Endothelial Cell Growth Medium 1 x 500 mL PM211500 10497-410 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human PrimaPure™ Endothelial Cell Growth Supplement 1 x 15 mL PM211GS 10497-676 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty