Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tronex Healthcare Fluid-Resistant, Spunbond, Full-Length Lab Coat with Lapel Collar, Knitted Cuffs, & Pockets, Tronex®

Supplier: Tronex
594035W 594045W 594020W 594010W 594040W 594030W
Designed to provide reliable protection and comfort, this meticulously crafted full-length lab coat is ideal for low to moderate fluid-exposure settings.

  • Constructed from a soft, breathable, fluid-resistant nonwoven spunbond material for excellent protection
  • Notch lapel collar and knitted cuffs for comfort
  • One hip pocket and one left chest pocket for convenience
  • Four sturdy front snaps for full closure
  • 100% latex-free

Certifications: Manufacturing certified ISO 9001 for Quality Management Systems. Manufacturing certified ISO 13485 for Quality Management Systems for Medical Devices.

Packaging: Lab Coats are packaged 5 per bag, 6 bags per case.



 

ORDER

Expand All / Collapse All
Clothing Size Color Supplier No. VWR Catalog Number Unit Price Quantity
S White 594010W 75818-186 Case of 30 Retrieving
M White 594020W 75818-188 Case of 30 Retrieving
L White 594030W 75818-190 Case of 30 Retrieving
XL White 594035W 75818-178 Case of 30 Retrieving
XXL White 594040W 75818-180 Case of 30 Retrieving
3XL White 594045W 75818-182 Case of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty