Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Human Dermal Fibroblast Cell Culture Kit (HDF), Genlantis

Supplier: Genlantis
PH10605NK PH10605N PH10605A PH10605FK PH10605F PH10605AK
The PrimaPure™ system has been developed for the acquisition and growth of primary cells which are derived from the dermis of normal human neonatal foreskin or adult skin. Fibroblasts are found in all connective tissues, and they synthesize and secrete extracellular matrix proteins under cell culture conditions.

  • Cryopreserved at the end of primary culture
  • Can be cultured and propagated at least 16 population doublings
  • Single donors to allow variability comparisons
  • Tested negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi

They are well established system for in vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Fibroblasts grown in a biodegradable mesh have been used as a living dermal replacement.

Delivery information: Each kit contains an ampoule of cryopreserved HDF, 500 mL of HDF Cell Growth Medium, and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Human Dermal PrimaPure™: Normal Adult Fibroblast Cells >500,000 Cryopreserved PH10605A 10497-634 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Dermal PrimaPure™: Normal Fetal Fibroblast Cells >500,000 Cryopreserved PH10605F 10497-638 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Dermal PrimaPure™: Normal Neonatal Fibroblast Cells >500,000 Cryopreserved PH10605N 10497-642 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Dermal PrimaPure™: Normal Adult Fibroblast, Complete System Kit - PH10605AK 10497-636 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Dermal PrimaPure™: Normal Fetal Fibroblast, Complete System Kit - PH10605FK 10497-640 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Dermal PrimaPure™: Normal Neonatal Fibroblast, Complete System Kit - PH10605NK 10497-644 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty