Model 409 Texture-Shield® Protective Lab Coat, Fashion Seal Healthcare®
Supplier: Superior Uniform Group
409-L 409-XL 409-S 409-XS 409-2XL 409-3XL
75819-464EA 100.85 USD
75819-464 75819-460 75819-470 75819-458 75819-466 75819-468
Model 409 Texture-Shield® Protective Lab Coat, Fashion Seal Healthcare®
Workwear
The Model 409 Texture-Shield® Protective Lab Coat is constructed entirely of Texture Shield® material
These white protective coats feature a concealed placket front with snap closures, a throat-guard collar with “hook and loop” closure for improved neck protection, two lower pockets, a back vent, and knit cuffs.