PrimaPure™, Human Epidermal Melanocytes Cell Culture Kit (HEM), Genlantis

Supplier: Genlantis
PH10405AK PH10405A
10497-632EA 1224.01 USD
10497-632 10497-630
PrimaPure™, Human Epidermal Melanocytes Cell Culture Kit (HEM), Genlantis
Cells
The PrimaPure™ system has been developed for the acquisition and growth of primary cells isolated from normal human neonatal foreskin

  • Cryopreserved at second passage
  • Can be cultured and propagated at least 12 population doublings
  • Single donors to allow variability comparisons
  • Tested negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

These epidermal melanocytes produce melanin, thus a useful cell model in the study of hyperpigmentation through accentuated melanocyte proliferation and differentiation as well as progression of melanocytic neoplasia.

Delivery information: Kit contains an ampoule of cryopreserved HEM, 500 mL of Melanocyte Growth Medium and a Subculture Reagent Kit (including 100 mL each of Hank's Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR