Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Human Epidermal Keratinocytes Cell Culture Kit (HEK), Genlantis

Supplier: Genlantis
PH10205AK PH10205NK PH10205N PH10205A PH10205FK PH10205F
The PrimaPure™ system has been developed for the acquisition and growth of primary cells which are isolated from normal human neonatal foreskin or adult skin.

  • Cryopreserved at the end of primary culture
  • Can be cultured and propagated at least 16 population doublings
  • Tested negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Single donors to allow variability comparisons

This kit is a suitable in vitro model for dermal toxicology tests in screening new skin products and for developing dermal skin substitutes

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Also used in numerous applications, including investigations of human epidermal development, differentiation and cellular aspects of skin diseases.

Delivery information: Each kit contains an ampoule of cryopreserved HEK, 500 mL of Keratinocyte Serum-Free Growth Medium, and a Subculture Reagent Kit (including 100 mL each of Hank's Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Human Skin PrimaPure™: Normal Adult Epidermal Keratinocyte Cells >500,000 Cryopreserved PH10205A 10497-618 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Skin PrimaPure™: Normal Fetal Epidermal Keratinocyte Cells >500,000 Cryopreserved PH10205F 10497-622 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Skin PrimaPure™: Normal Neonatal Epidermal Keratinocyte Cells >500,000 Cryopreserved PH10205N 10497-626 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Skin PrimaPure™: Normal Adult Epidermal Keratinocyte, Complete System Kit - PH10205AK 10497-620 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Skin PrimaPure™: Normal Fetal Epidermal Keratinocyte, Complete System Kit - PH10205FK 10497-624 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Human Skin PrimaPure™: Normal Neonatal Epidermal Keratinocyte, Complete System Kit - PH10205NK 10497-628 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty