PrimaPure™, Human Dermal Microvascular Endothelial Cell Culture Kit (HMVEC), Genlantis

Supplier: Genlantis
PH10005NK PH10005A PH10005AK PH10005N
10497-616EA 1754.29 USD
10497-616 10497-614 10497-612 10497-610
PrimaPure™, Human Dermal Microvascular Endothelial Cell Culture Kit (HMVEC), Genlantis
Cells
The PrimaPure™ system has been developed for the acquisition and growth of primary cells isolated from normal human neonatal foreskin or adult skin capillaries. They can be cultured and propagated at least 16 population doublings. They are responsive to cytokine stimulation in the expression of cell adhesion molecules. There is growing evidence of differences in physiology and pharmacology between microvascular endothelial cells and macrovascular endothelial cells.

  • Tested positive for CD36 expression
  • Von Willebrand factor/factor VIII (vWF) positive
  • Negative for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Cryopreserved at second passage
  • Single donors to allow variability comparisons

HMVEC is an organ-specific endothelial cell system for the study of angiogenesis, tumor metastasis, wound healing, inflammation, macromolecule transport, and drug metabolism.

Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.

Delivery information: Kit contains an ampoule of cryopreserved HMVEC, 500 mL of HMVEC Cell Growth Medium, a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, trypsin/EDTA, and trypsin neutralizing solution), two Extracellular Matrix Protein Coated T-25 flasks and Attachment Factor Solution.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR