SmartCells™ High Efficiency Chemically Competent E. coli, Genlantis
Supplier: Genlantis
C101020 C101120
10497-296EA 552.18 USD
10497-296 10497-294
SmartCells™ High Efficiency Chemically Competent E. coli, Genlantis
Competent Cells
SmartCells™ chemically competent E.coli are prepared by a unique procedure to warrant the highest and most robust transformation performance under diverse conditions.
  • No dilution or purification required
  • Carry genotypes suitable for most cloning needs
Ideal for blue/white selection, generation of plasmid vector based libraries or gene banks, and has the ability to be transformed efficiently with large plasmids. SmartCells™ F’ is also available for use with M13 cloning vectors.
Delivery information: Includes SmartCells™ chemically competent E.coli, SOC medium (6 mL) and pUC19 positive control plasmid (20 µL, 10pg/µL).