Human [Pro19]-beta-Amyloid (1-42)

Supplier: AnaSpec
AS-65178
103009-022EA 704.46 USD
103009-022
Human [Pro19]-beta-Amyloid (1-42)
Proteins and Peptides
This peptide is beta-amyloid (1-42) with substitution of Phe19 to Pro. This substitution inhibits amyloid fibril formation, which is implicated in neurodegenerative diseases such as Alzheimer’s.
Sequence: DAEFRHDSGYEVHHQKLVFFAENVGSNKGAIIGLMVGGVVIA
MW: 4513.1 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR