Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 15D Digital Homogenizer with Brushless Motor

Supplier: VWR International
PL-03-21002P PL-02-43200 PL-99-03302P PL-02-20115 PL-03-21003P PL-02-20200 PL-02-05075 PL-03-11703P PL-02-10115 PL-02-43340 PL-02-07095 PL-03-11004P PL-03-11037P PL-03-11005P PL-02-30200 VWR-01-01370 PL-02-59200 PL-02-37200 PL-02-37340
The VWR 15D homogenizer can be used for disruption of tissues samples in biotechnology, sample preparation in medicine, enzymatic treatment in the food industry as well as various applications in pharmaceutical, cosmetic, paint, petro chemical, and other industrial applications.

  • Quiet maintenance free drive
  • Digital display and variable speed control
  • Perfect for clean room applications
  • 0.05ml - 1.5 L volume range and viscosity up to 5,000 mPas
  • VWR® Generator probe sold separately

The VWR 15D Homogenizer's brushless motor guarantees numerous years of maintenance free operation as it has no carbon brushes, so it runs cleaner and quieter

It can process samples from 0.05ml to 1.5L (higher sample volumes dependent on viscosity).

This allows end-users to operate without ear plugs or without the annoyance of louder noise heard from brush type homogenizers. The VWR15D homogenizer's brushless design makes it ideal for processing volatile or flammable organic compounds and buffer too.

Certifications: CE marked.

Accessories information: Generator probes must be purchased with homogenizer units.

Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR15D Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes).



 

ORDER

Expand All / Collapse All

Homogenizer
Description Electrical Volume Range Speed Range VWR Catalog Number Unit Price Quantity
VWR15D Brushless Digital Homogenizer 120V 0.05ml - 1.5 L 0 - 16,500 RPM 75800-450 Each Retrieving

Generator Probes
Description Details For Model VWR Catalog Number Unit Price Quantity
VWR® Flat-Bottom Generator Probe, 5 x 75 mm For 0.5–2 mL Tubes 200; 250; 15D; 25D 10032-562 Each Retrieving
VWR® Saw-Tooth Generator Probe, 7 x 95 mm For 1.5–10 mL Tubes 200; 250; 15D; 25D 10032-566 Each Retrieving
VWR® Saw-Tooth Generator Probe, 10 x 115 mm For 10–50 mL Tubes 200; 250; 300D; 400D; 15D; 25D 10032-568 Each Retrieving
VWR® Saw-Tooth Generator Probe, 20 x 115 mm For 50 mL Tubes and Small Beakers 200; 250; 300D; 400D; 15D; 25D 10032-570 Each Retrieving
VWR® Saw-Tooth Generator Probe, 20 x 200 mm For Medium to Large Beakers 250; 300D; 400D; 15D; 25D 10032-572 Each Retrieving
VWR® Open-Slotted Generator Probe, 30 x 200 mm For Large Beakers and Containers 250; 300D; 400D; 25D 10032-574 Each Retrieving
VWR® Open-Slotted Generator Probe, 37 x 200 mm For Large Beakers and Containers 300D; 400D 10032-576 Each Retrieving
VWR® Open-Slotted Generator Probe, 37 x 340 mm For Large Containers 400DEL 10032-578 Each Retrieving
VWR® Open-Slotted Generator Probe, 43 x 200 mm For Large Beakers and Containers 300D; 400D 10032-580 Each Retrieving
VWR® Open-Slotted Generator Probe, 43 x 340 mm For Large Containers 400DEL 10032-582 Each Retrieving
VWR® Open-Slotted Generator Probe, 59 x 200 mm For Large Beakers and Containers 300D; 400D 10032-584 Each Retrieving

Generator Probe Replacement Components
Description For Model VWR Catalog Number Unit Price Quantity
Replacement Bearing, GenPk. Pack of 6 200; 250; 300D; 400D; 15D; 25D 75839-976 Pack of 6 Retrieving
Replacement Lower Flanged Bearing, 10 mm, PTFE, Pack of 2 200; 250; 300D; 400D; 15D; 25D 75840-002 Pack of 2 Retrieving
Replacement Lower Flanged Bearing, 20 mm, PTFE, Pack of 2 200; 250; 300D; 400D; 15D; 25D 75840-004 Pack of 2 Retrieving
Replacement Lower Flanged Bearing, 30 mm, PTFE, Pack of 2 250; 300D; 400D; 15D; 25D 75839-974 Pack of 2 Retrieving
Replacement Lower Sleeve Bearing, 5 mm, PTFE, Pack of 2 200; 250; 300D; 400D; 15D; 25D 75839-978 Pack of 2 Retrieving
Replacement Lower Sleeve Bearing, 7 mm, PTFE, Pack of 2 200; 250; 300D; 400D; 15D; 25D 75839-980 Pack of 2 Retrieving
Replacement Upper Washer, PTFE, Pack of 5 200; 250; 300D; 400D; 15D; 25D 75839-982 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Weight (kg) 7.71 kg (17 lbs.)
Dimensions 24.13 x 17.78 x 63.5 cm (9.5 x 7 x 25")
Power rating 150W
Noise Rating 55 dB

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details For Model VWR Catalog Number Unit Price Quantity
Small Stand Assembly for VWR® 250 Homogenizer 4.5 kg (10 lbs.), 15.2 x 15.2 x 60.9 cm (6 x 6 x 24") 250 10032-554 Each Retrieving
Stand Assembly for VWR® 200 Homogenizer 4.5 kg (10 lbs.), 15.2 x 15.2 x 60.9 cm (6 x 6 x 24") 200 10032-346 Each Retrieving
Stand Assembly for VWR® 250 Homogenizer 4.5 kg (10 lbs.), 30.5 x 35.5 x 60.9 cm (12 x 14 x 24") 250 10032-348 Each Retrieving
VWR® GenPack Adapter Required for GenPack 200; 250; 300D; 400D; 15D; 25D 10032-558 Each Retrieving
VWR® GenPack Generator Probes, 12-Pack For 1.5–15 mL Tubes 200; 250; 300D; 400D; 15D; 25D 10032-560 Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty