Trail Receptor 2, CD262, Human ELISA, BioVendor
Supplier: BioVendor
RGP023R
102552-630EA 565 USD
102552-630
Trail Receptor 2, CD262, Human ELISA, BioVendor
Assays ELISAs
Trail R2 transduce an apoptosis signal
  • Calibration Range: 31.25–1000 pg/mL
  • Limit of Detection: 6 pg/mL
  • Cell culture lysate, Cell culture supernatant, Serum: 100 uL/well
Human Trail R1 (R2) is a TNF receptor , which is a receptor for Trail (APO2 ligand). Trail R1 and R2 are involved in apoptosis. In the Trail receptor family, Trail R1, but also Trail R2 transduce an apoptosis signal, whereas Trail R3 and Trail R4 antagonize TRAIL-induced apoptosis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.