Would you like to log in again?
Fibroblast Growth Factor 21, FGF21, Human ELISA, BioVendor
Supplier: BioVendor
RD191108200R
FGF21 has been characterised as a potent metabolic regulator.
  • Calibration Range: 30-1920 pg/mL
  • Limit of Detection: 7 pg/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 75 uL/well
The fibroblast growth factor family (FGFs) are a family of more than 20 small (17–26 kDa) secreted peptides. The initial characterisation of these proteins focused on their ability to stimulate fibroblast proliferation through FGF receptors (FGFRs). Members of FGFs family play important roles in defining and regulating the development and function of endocrine tissues as well as modulating various metabolic processes. A recently described member of FGFs family, FGF-21, also called Fibroblast growth factor 21 precursor and UNQ3115/PRO10196, has been characterised as a potent metabolic regulator. FGF-21 is preferentially expressed in liver and regulates glucose uptake in human fat cells. Moreover, therapeutic administration of FGF-21 decreased plasma glucose levels and triglycerides to near normal levels in multiple mouse models of type 2 diabetes. Short-term treatment of normal or db/db mice with FGF-21 lowered plasma levels of insulin and improved glucose clearance compared with vehicle after oral glucose tolerance testing. Constant infusion of FGF-21 for 8 weeks in db/db mice nearly normalized fed blood glucose levels and increased plasma insulin levels. When administrated daily for 6 weeks to diabetic rhesus monkeys, FGF-21 caused dramatic decline in fasting plasma glucose, fructosamine, triglicerides, insulin, and glucagon. FGF-21 administration also led to significant improvements in lipoprotein profiles, including lowering of low-density lipoprotein cholesterol and raising of high-density lipoprotein cholesterol as well as beneficial changes in the circulating levels of several cardiovascular risk factors. FGF-21, when overexpressed, protected animals from diet-induced obesity. These results define a functional role for FGF-21 in vivo and provide evidence that FGF-21 can lower glucose and triglyceride levels in diabetic animals. In contrast to several members of the FGF family which may induce therapeutically undesirable in vivo proliferation of various cell types, a recent study demonstrated that FGF-21 did not induce mitogenicity, hypoglycemia or weight gain at any dose tested in diabetic or healthy animals or when overexpressed in transgenic mice. Thus, FGF-21 appears to have considerable potential for the treatment of diabetes mellitus.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Fibroblast Growth Factor 21 (FGF21) Human ELISA 1 kit RUO RD191108200R 102552-224 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty