Would you like to log in again?
Prolactin Rat ELISA, BioVendor
Supplier: BioVendor
RTC006R
Rat prolactin differs from human prolactin at about 50 percent of all residues.
  • Calibration Range: 5-80 ng/mL
  • Limit of Detection: 0.6 ng/mL
  • Serum: 25 uL/well
Rat prolactin (rPRL) is a single-chain polypeptide hormone of the rat anterior pituitary with a molecule mass of approximately 23,000. Prolactin from different species exhibits significant variations in the amino acid sequence. Rat prolactin differs from human prolactin at about 50 percent of all residues. The secretion of rPRL from the pituitary is inhibited by hypothalamic prolactin-inhibitory factor (PIF). Although dopamine was long thought to be this PIF molecule, today it seems that there is a special peptide with prolactin-inhibiting activities. The release of prolactin is certainly stimulated by different peptides, particularly thyrotropin releasing hormone (TRH) and vasoactive intestinal peptide (VIP). There is also evidence that rat posterior pituitary lobe contains a special prolactin releasing hormone. The most important role of prolactin is stimulation of mammary gland growth and lactation. During pregnancy, blood prolactin levels climb, but the increases can differ enormously between rats. High prolactin levels are observed during lactation. Prolactin has a wide variety of other physiological actions, for example on the ovary. In the rat, prolactin has a luteotrophic effect which is not seen in many other species. Furthermore, prolactin is a stress hormone. In rats, as in humans, prolactin exhibits a sleep-related diurnal variation. Peak values are seen in the late afternoon and nadir values in the morning. Because of the variety of its actions, prolactin is one of the preferred hormones to monitor when testing the influence of new therapeutic agents and drugs on the endocrine system in the rat.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
Prolactin Rat ELISA 1 kit RUO RTC006R 101076-644 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty